Stampa…

Anti-VIM Mouse Monoclonal Antibody (Purified) [clone: LN-6]

Fornitore: Biotium

BNUB0060-500 BNUB0060-100
BNUB0060-500EA 483 EUR
BNUB0060-500 BNUB0060-100
Anti-VIM Mouse Monoclonal Antibody (Purified) [clone: LN-6]
Anticorpi
This MAb reacts with a 58 kDa protein identified as vimentin. It reacts with a non-hematopoietic epitope of vimentin and shows no cross-reaction with other closely related intermediate filament proteins (IFP's) such as desmin, keratin, neurofilament, and glial fibrillary acid protein.Vimentin is ubiquitously expressed in mesenchymal cells such as fibroblasts, smooth muscle cells, and endothelium. Antibody against vimentin is useful as part of an antibody panel for differential diagnosis of tumors of unknown origin.Ab-2 does not react with leukocyte common antigen-positive tissues such as lymphomas and leukemias.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: LN-6
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Cat, Cow, Human, Mouse, Pig, Rabbit, Rat, Sheep
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR