Stampa…
Linseed oil (raw), clear yellow liquid
Fornitore: MP Biomedicals
Attenzione
0296012201 0296012205
ICNA0296012201EA 50.8 EUR
ICNA0296012201 ICNA0296012205
Linseed oil (raw), clear yellow liquid
Linseed oil
  • Iodine Value: Approximately 170
  • Saponification Value: Approximately 187 - 195 (Lit.)
MDL Number: MFCD00131507
CAS Number: 8001-26-1