Linseed oil (raw), clear yellow liquid
Fornitore: MP Biomedicals
|
Attenzione
|
0296012201
0296012205
ICNA0296012201EA
50.8
EUR
ICNA0296012201
ICNA0296012205
Linseed oil (raw), clear yellow liquid
Linseed oil
- Iodine Value: Approximately 170
- Saponification Value: Approximately 187 - 195 (Lit.)
MDL Number:
MFCD00131507 CAS Number: 8001-26-1 |