Stampa…

Hypotaurine, white prills

Fornitore: MP Biomedicals
Attenzione

0215971180 0215971183
ICNA0215971180EA 62.4 EUR
ICNA0215971180 ICNA0215971183
Hypotaurine, white prills
Hypotaurine

Hypotaurine is a Non-specific, endogenous inhibitory glycine receptor agonist.


An antioxidant that binds hydroxyl radicals. Endogenous inhibitory neuromodulator that is blocked by strychnine and picrotoxin. Inhibits the Na2+-dependent transport of GABA and β-alanine. It is a degradation product of cysteine and a precursor of taurine.


Soluble in water (100 mM - colorless to faint yellow solution, 100 mg/ml).

Formula: C₂H₇NO₂S
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00038197
CAS Number: 300-84-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR