9-cis-Retinoic acid ≥98%, in polvere
Fornitore: MP Biomedicals
|
|
Synonyms:
9-cis-RA, NSC-659772, Alitretinoin, Panretin, (2E,4E,6Z,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
ICNA02158725
9-cis-Retinoic acid inhibits cell proliferation and induces cell differentiation.
9-cis-Retinoic acid is a high affinity ligand for the retinoid X receptor (RXR), it destabilize Vitamin D receptor RXR heterodimer-DNA complexes. It is naturally produced via isomerization of all-trans retinoic acid.
Soluble in DMSO (25 mg/ml), acetone (25 mg/ml) and ethanol (25 mg/ml). To solubilise the product in ethanol at 25 mg/ml the solution needs to be warmed to 50 °C. Store solutions aliquoted at -20 °C in the dark for up to 3 months.
Formula:
C₂₀H₂₈O₂ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00270072 CAS Number: 5300-03-8 |
Risultati del test di specificazione
Identity Test | Passes |
Purity | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...