Stampa…

N,N'-Octamethylenebis(2,2-dichloroacetamide), off-white powder

Fornitore: MP Biomedicals
Attenzione

Synonyms: N,N'-(Octane-1,8-diyl)bis(2,2-dichloroacetamide), WIN18446

0215805083 0215805091
ICNA0215805083EA 110 EUR
ICNA0215805083 ICNA0215805091
N,N'-Octamethylenebis(2,2-dichloroacetamide), off-white powder
N,N'-Octamethylenebis(2,2-dichloroacetamide)

N,N'-Octamethylenebis(2,2-dichloroacetamide) is an inhibitor of aldehyde dehydrogenase, ALDH1A2.

Formula: C₁₂H₂₀Cl₄N₂O₂
Peso molecolare: 366,104 g/mol
Punto di fusione: 128,0…128.4 °C (lett.)
MDL Number: MFCD00000841
CAS Number: 1477-57-2

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR