Stampa…

Ethoxyquin ≥90%, viscous liquid

Fornitore: MP Biomedicals

Synonyms: 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin

0215796380 0215796383 0215796391
ICNA0215796380EA 87.7 EUR
ICNA0215796380 ICNA0215796383 ICNA0215796391
Ethoxyquin ≥90%, viscous liquid
Ethoxyquin

Ethoxyquin is a quinoline-based antioxidant.


Ethoxyquin is an antioxidant which inhibits chemically induced carcinogenesis, where it exerts its chemopreventive action by inducing glutathione synthetase and the phase II drug metabolizing enzyme, glutathione S-transferase (GST) and an antidegradation agent for rubber.

Formula: C₁₄H₁₉NO
MDL Number: MFCD00023883
CAS Number: 91-53-2

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity ≥90%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR