Stampa…

Lactulose ~98%, white powder

Fornitore: MP Biomedicals

0215393280 0215393205 0215393225
ICNA0215393280EA 461 EUR
ICNA0215393280 ICNA0215393205 ICNA0215393225
Lactulose ~98%, white powder
Lactulose

Lactulose is a synthetic, non-digestible sugar used in the treatment of chronic constipation and hepatic encephalopathy, a complication of liver disease. It is a disaccharide (double-sugar) formed from one molecule each of the simple sugars (monosaccharides) fructose and galactose.


Lactulose is used as a test of small intestine bacterial overgrowth (SIBO). Lactulose is used in the treatment of chronic constipation. It is useful in treating the hyperammonemia caused by hepatic encephalopathy, because it helps "draw out" ammonia from the body. Lactulose is also a food ingredient, better known as galactofructose, with sweet taste and offering beneficial health benefits on digestive health.


Soluble in 5% of water (colorless)

Formula: C₁₂H₂₂O₁₁
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00151469
CAS Number: 4618-18-2

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity ~98%
FTIR Conforms to Reference
Moisture Content 3,0%
Optical Rotation -45 to -52° (c=5, water, 24 hrs)
pH (1% Aqueous solution) 5,5 - 6,5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR