Stampa…

Cortisone, in polvere

Fornitore: MP Biomedicals

Synonyms: 17alpha,21-Dihydroxypregn-4-ene-3,11,20-trione

02190172.2 0219017201 0219017205
ICNA02190172.2EA 35.7 EUR
ICNA02190172.2 ICNA0219017201 ICNA0219017205
Cortisone, in polvere
Cortisone

Cortisone is a glucocorticoid; a corticosterone analog that has approximately twice the anti-inflammatory potency as corticosterone but much lower Na2+ retention potency.


Soluble in acetone (10 mg/ml - clear, colorless solution).

Formula: C₂₁H₂₈O₅
Peso molecolare: 360.44 g/mol
Punto di fusione: 223…228 °C (dec., lit.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003610
CAS Number: 53-06-5

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR