Stampa…

Tetrabutylammonium dihydrogen phosphate ∼ 97%, in polvere

Fornitore: MP Biomedicals

Synonyms: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

0219551301 0219551305
ICNA0219551301EA 60.4 EUR
ICNA0219551301 ICNA0219551305
Tetrabutylammonium dihydrogen phosphate ∼ 97%, in polvere
Tetrabutylammonium dihydrogen phosphate

Ion-associating reagent for HPLC


  • Ion-associating reagent for HPLC


Soluble in water (clear to hazy).


Attenzione: Research Use Only (RUO).

Formula: C₁₆H₃₈NO₄P
Peso molecolare: 339,46 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00064526
CAS Number: 5574-97-0

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity ~97%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR