Stampa…

Sodio diidrogeno fosfato monoidrato 98.0-102.0%, white powder ACS

Fornitore: MP Biomedicals

02191442.1 02191442.5 0219144201
ICNA02191442.1EA 32.9 EUR
ICNA02191442.1 ICNA02191442.5 ICNA0219144201
Sodio diidrogeno fosfato monoidrato 98.0-102.0%, white powder ACS
Sodio diidrogeno fosfato monoidrato

Sodium phosphate monobasic monohydrate is useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.


Freely soluble in water; insoluble in ethanol.


Attenzione: Research Use Only (RUO).

Formula: NaH₂PO₄·H₂O
MDL Number: MFCD00149208
CAS Number: 10049-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity 98.0% - 102.0%
pH 4,1 - 4,5 (5% aq soln)
Insoluble Matter ≤0,01%
Chloride ≤5 ppm
Sulphate ≤0,003%
Calcium ≤0,005%
Potassium ≤0,01%
Heavy Metals (as Pb) ≤0,001%
Iron ≤0,001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR