Cyproheptadine hydrochloride sesquihydrate, white powder
Fornitore: MP Biomedicals
Synonyms:
Cyproheptadine hydrochloride, 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methylpiperidine hydrochloride
Cyproheptadine is a Ca2+ channel blocker; 5-HT2 serotonin receptor antagonist; antihistamine, antipruritic. It is a migraine prophylactic.
- 5-HT2/5-HT1C serotonin receptor antagonist
- H1 histamine receptor antagonist
- Antipruritic.
5-HT2/5-HT1C serotonin receptor antagonist; H1 histamine receptor antagonist; antipruritic.
Soluble in water, methanol, alcohol or ethanol.
Formula:
C₂₁H₂₁N·HCl Peso molecolare: 350,88 g/mol |
MDL Number:
MFCD00012538 CAS Number: 41354-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...