Stampa…

Triolein ≥95%, light yellow, clear liquid

Fornitore: MP Biomedicals

Synonyms: Glyceryl trioleate, Glycerine trioleate, Glycerol Trioleate

0210312201 0210312205 0210312210
ICNA0210312201EA 79.8 EUR
ICNA0210312201 ICNA0210312205 ICNA0210312210
Triolein ≥95%, light yellow, clear liquid
Triolein

Triolein is a symmetrical triglyceride.


Triolein is used as a substrate for microbial lipases.


Soluble in chloroform, ether, carbon tetrachloride; slightly soluble in alcohol; practically insoluble in water.


Attenzione: Research Use Only (RUO).

Formula: C₅₇H₁₀₄O₆
Peso molecolare: 885,45 g/mol
Punto di ebollizione: 235…240 °C (18 mmHg)) (lett.)
Punto di fusione: –4 °C (lett.)
Densità: 0,91 g/mL (lett.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00137563
CAS Number: 122-32-7

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity ≥95%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR