Stampa…

Octenidine dihydrochloride 98%

Fornitore: Thermo Fisher Scientific

Synonyms: 1,1'-(Decane-1,10-diyl)bis(N-octylpyridin-4(1H)-imine) dihydrochloride

H62257.03 H62257.06
H62257.03EA 46.8 EUR
H62257.03 H62257.06
Octenidine dihydrochloride 98%
Octenidine dihydrochloride

Octenidine dihydrochloride is used as a antiseptic for skin, mucous Membranes and wounds.

Formula: C₃₆H₆₂N₄
Punto di fusione: 214 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01938808
CAS Number: 70775-75-6
EINECS: 274-861-8
Merck Index: 14,6754

Visualizza i codici

Risultati del test di specificazione

Synonyms N,N'-[Decane-1,10-diyldi-1(4H)-pyridyl-4-ylidene]bis(octylammonium) dichloride
Solubility Soluble in dimethyl sulfoxide and methanol
Assay (HPLC) 97,5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR