Stampa…

Phosphatidyl-L-serine ~98%

Fornitore: MP Biomedicals
Pericolo

0210096910 0210096950
ICNA0210096910EA 266 EUR
ICNA0210096910 ICNA0210096950
Phosphatidyl-L-serine ~98%
Phosphatidyl-L-serine

A slowly metabolised structural phospholipid found mainly in gray matter.


Phosphatidyl Serine has been reported to have positive effects on mental function and mood. It has been prescribed to those suffering from age associated cognitive decline. It has also been suggested that Phosphatidyl Serine could help improve aerobic endurance by delaying the onset of fatigue.


Solutions exposed to room temperature will decompose about 0,5% per day.


Attenzione: Research Use Only (RUO).

Formula: C₄₂H₈₂NO₁₀P
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00135635
CAS Number: 51446-62-9

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity ~98%
Source Bovine brain
Concentration 10 - 40 mg/mL
Supplied in Chloroform
Storage Store at - 0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR