Stampa…

5-Ethylphenazinium ethyl sulphate ∼95%, in polvere

Fornitore: MP Biomedicals
Attenzione

Synonyms: PES, Phenazine ethosulphate

02100956.5 0210095601 0210095605
ICNA02100956.5EA 57.6 EUR
ICNA02100956.5 ICNA0210095601 ICNA0210095605
5-Ethylphenazinium ethyl sulphate ∼95%, in polvere
5-Ethylphenazinium ethyl sulphate

Phenazine ethosulfate is an intermediate which is used to detect nitric oxide reducatase activity in concert with ascorbic acid.


Soluble in water (10 mg/ml - clear, yellow solution) or ethanol (50 mg/ml - clear, dark yellow to orange solution).


Attenzione: Research Use Only (RUO).

Formula: C₁₆H₁₈N₂O₄S
Peso molecolare: 334,4 g/mol
Temperatura di stoccaggio: Congelamento
CAS Number: 10510-77-7

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity ~95%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR