Stampa…
HPLC columns, Polaris™ amide C18
Fornitore: VARIAN
A2006050X020 A2006050X046 A2006150X046 A2006250X046 A2007030X030 A2007050X020 A2007050X030 A2007050X046 A2007150X046 A2007250X046 A2006030X020 A2006100X020 A2006100X030 A2006150X020 A2006150X030 A2006250X020 A2006250X030 A2006250X100 A2007050X021 A2007050X032 A2007050X100 A2007100X020 A2007100X021 A2007100X030 A2007100X046 A2007150X020 A2007150X021 A2007150X030 A2007150X032 A2007250X020 A2007250X030 A2008250X100
VARIA2006050X020EA 896 EUR
VARIA2006050X020 VARIA2006050X046 VARIA2006150X046 VARIA2006250X046 VARIA2007030X030 VARIA2007050X020 VARIA2007050X030 VARIA2007050X046 VARIA2007150X046 VARIA2007250X046 VARIA2006030X020 VARIA2006100X020 VARIA2006100X030 VARIA2006150X020 VARIA2006150X030 VARIA2006250X020 VARIA2006250X030 VARIA2006250X100 VARIA2007050X021 VARIA2007050X032 VARIA2007050X100 VARIA2007100X020 VARIA2007100X021 VARIA2007100X030 VARIA2007100X046 VARIA2007150X020 VARIA2007150X021 VARIA2007150X030 VARIA2007150X032 VARIA2007250X020 VARIA2007250X030 VARIA2008250X100
HPLC columns, Polaris™ amide C18
Colonne cromatografiche
Polaris™ amide C18 has the greatest polarity of the Polaris™ C18 family. The amide polar groups offer a different selectivity from either the C18-A or C18-ether phases. The surface has been covalently modified with alkyl amide groups and endcapped.
  • Hydrophobic H-bonding
  • Spherical particles with pore size 180 Å