Stampa…
Agarose, MetaPhor™
Fornitore: LONZA
50181 50180 50184
733-1211EA 302 EUR
733-1211 733-1210 733-1212
Agarose, MetaPhor™
Gel elettroforetici Agarosio e Acrilammide
Intermediate melting point agarose ideal for resolution of DNA fragments, PCR and RT-PCR products, differing in size by 2%, in the range of 20 to 800 bp, by submarine gel electrophoresis.
MetaPhor™ agarose gel is ideal for resolving AMPFLPs, STRs, and tri- and tetranucleotide repeats.