Stampa…
Nitrile gloves, TouchNTuff® 93-163 / 93-263
Fornitore: Ansell
93-163/6.5-7 93-163/10.5-11 93-163/7.5-8 93-163/8.5-9 93-163/9.5-10 93-263/7.5-8 93-263/6.5-7 93-263/9.5-10 93-263/10.5-11 93-263/8.5-9
ANSE93-163/6.5-7EA 61.4 EUR
ANSE93-163/6.5-7 ANSE93-163/10.5-11 ANSE93-163/7.5-8 ANSE93-163/8.5-9 ANSE93-163/9.5-10 ANSE93-263/7.5-8 ANSE93-263/6.5-7 ANSE93-263/9.5-10 ANSE93-263/10.5-11 ANSE93-263/8.5-9
Nitrile gloves, TouchNTuff® 93-163 / 93-263
Guanti
These disposable gloves feature a special nitrile formulation for low hand fatigue.
  • Ambidextrous, easy donning
  • Textured finish, light and resistant
  • Good puncture resistance
  • Flexible and comfortable offering excellent dexterity
  • Beaded cuff for an easy donning and offering good tear resistance
Typical applications include: Chemical handling, glass manufacturing, handling of cytostatics, laboratory analysis, light assembly tasks, paint and spray shops, pharmaceuticals and the printing industry.
Certificazioni: EN ISO 374-1:2016 Type B (JKT), EN ISO 374More Product Information
Visualizza i codici
Specifiche
Modello 93-163 93-263 93-263
Ambidestro/mano specifico ambidestro ambidestro ambidestro
Environmentally Preferable
Monouso/riutilizzabile monouso monouso monouso
Lattice/senza lattice Senza lattice Senza lattice Senza lattice
Talcati/non talcati Non talcati Non talcati Non talcati
Finitura Textured Textured Textured
Tipo polsino Cuff_Style-21391099|Cuff_Style-21391103 Cuff_Style-21391099|Cuff_Style-21391103 Cuff_Style-21391103|Cuff_Style-21391099
Norma EN EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 VIRUS, EN 421 EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 VIRUS, EN 421 EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5:2016 VIRUS, EN 421
Approvati per il contatto con gli alimenti + + +