Powder Pillows, nickel reagents
Fornitore: Hach
212368
212468
705-0094EA
47.4
EUR
705-0094
705-0095
Powder Pillows, nickel reagents
Kit per analisi ambientali
USEPA accepted for reporting wastewater analysis (digestion required). Procedure is equivalent to Standard Method 3500-Ni D for wastewater.