Stampa…
Acidic cleaner and neutraliser, deconex® 26 PLUS
Fornitore: Borer Chemie
502300.00-KK5R 502300.00-K10R 502300.00-K25R
148-5265EA 149 EUR
148-5265 148-5266 DECO502300.00-K25R
Acidic cleaner and neutraliser, deconex® 26 PLUS
Soluzioni detergenti Detergenti e Detersivi per Superfici
Liquid, highly acidic cleaning concentrate und neutraliser for automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees consistent and error-free results
  • Neutralises carried over alkaline residues
  • Substantially reduces rinsing time and consumption of water
  • In combination with all alkaline deconex® cleaning products, can be also used as a precleaner to remove acid soluble deposits
  • Removes lime residues in laboratory washers
  • Ideal for an effective cleaning of animal cages, also suitable for use in laboratories in the dairy industry
  • EDTA-, NTA-, chlorine- and surfactant-free, no release of surfactants into waste water