Itraconazole 99%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Oriconazole
452870050
452870250
452870000
ACRO452870050EA
115
EUR
ACRO452870050
ACRO452870250
ACRO452870000
Itraconazole 99%
Itraconazole
Formula:
C₃₅H₃₈Cl₂N₈O₄ Peso molecolare: 705,64 g/mol Punto di fusione: 168…170 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00870168 CAS Number: 84625-61-6 |
Risultati del test di specificazione
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Melting point | 165 to 169 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...