Stampa…

Calcio acetato monoidrato ACS

Fornitore: Thermo Fisher Scientific

Synonyms: Calcium di(acetate) monohydrate, Calcium acetate hydrate, Acetic acid calcium salt monohydrate

446351000 446355000
ACRO446351000EA 55.9 EUR
ACRO446351000 ACRO446355000
Calcio acetato monoidrato ACS
Calcio acetato monoidrato
Formula: (H₃CCOO)₂Ca·1H₂O
Punto di fusione: 160 °C
Densità: 1,5 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00150019
CAS Number: 5743-26-0

Visualizza i codici

Risultati del test di specificazione

Appearance White Crystalline powder
Titration Complexometric ≥99.0 %
Alkalinity Passes test
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.01 %
Barium (Ba) ≤0.01 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.05 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.05 %
Heavy metals (as Pb) ≤0.005 %
Titrable acid ≤0.035 meq/g
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR