Acetoacetamide 97%
Fornitore: Thermo Fisher Scientific
Synonyms:
Acetylacetamide, 3-Oxobutanamide, 3-oxobutyramide
Formula:
C₄H₇NO₂ Punto di fusione: 53…56 °C Flash Pt: 113 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00025528 CAS Number: 5977-14-0 |
Risultati del test di specificazione
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 52 to 58 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...