Stampa…

D-Propargylglycine 98%, 98% ee

Fornitore: Thermo Scientific
Attenzione

Synonyms: H-D-Pra-OH, (R)-2-Aminopent-4-ynoic acid, (R)-alpha-Propargylglycine

441221000 441225000
ACRO441221000EA 119 EUR
ACRO441221000 ACRO441225000
D-Propargylglycine 98%, 98% ee
D-Propargylglycine
Formula: C₅H₇NO₂
Peso molecolare: 113,116 g/mol
Punto di fusione: >180 °C (dec.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00237407
CAS Number: 23235-03-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to light yellow Powder
Infrared spectrum Conforms
Assay 97.5 to 102.5 %
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=1 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR