Stampa…

Mannide monooleate

Fornitore: Thermo Scientific

Synonyms: 1-O-[(9Z)-9-Octadecenoyl]-D-glucopyranose, (Z)-dianhydro-D-mannitol, mono-9-octadecenoate, D-Glucopyranose, 1-O-[(9Z)-1-oxo-9-octadecen-1-yl]-

332130250 332131000
332130250.EA 70.6 EUR
332130250. 332131000.
Mannide monooleate
Mannide monooleate
Formula: C₂₄H₄₄O₇
Peso molecolare: 444,609 g/mol
Densità: 0,97 g/cm³
Flash Pt: >149 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03840565
CAS Number: 25339-93-9

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to brown Viscous liquid
Infrared spectrum Conforms
Water ≤0.2 %
Acid value ≤1 mg KOH/g
Hydroxyl value 90 to 100 mg KOH/g
Iodine value 70 to 78 g I2/100 g
Saponification value 164 to 172 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR