Bovine serum albumin (BSA)
Fornitore: Thermo Scientific
268130100
268131000
268135000
ACRO268130100EA
72.7
EUR
ACRO268130100
ACRO268131000
ACRO268135000
Bovine serum albumin (BSA)
Proteine e Peptidi
- Off-white to yellow-beige to amber Lyophilised powder
Loss on drying: =<5,0%
Albumin: >=98% (HPLC)
pH: 6,7 to 7,3 (10 % solution)
Protease: None detected
IgG: None detected
Chloride (Cl): =<50 meq/L (10% solution)
Sodium (Na): =<50 meq/L (10% solution)
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...