Stampa…

1,2-Dipalmitoyl-sn-glycero-3-phosphocholine 98%

Fornitore: Thermo Scientific

Synonyms: 2,3-Bis(palmitoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(palmitoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate, L-DPPC, 1,2-Dihexadecanoyl-sn-glycero-3-phosphocholine, L-α-Lecithin, Dipalmitoyl, L-alpha-Dipalmitoyl phosphatidylcholine

240575000 240570025 240571000
ACRO240575000EA 334 EUR
ACRO240575000 ACRO240570025 ACRO240571000
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine 98%
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine
Formula: C₄₀H₈₀NO₈P
Peso molecolare: 734,05 g/mol
Punto di fusione: 229…229,5 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00036903
CAS Number: 63-89-8

Visualizza i codici

Risultati del test di specificazione

Appearance White Powder
Infrared spectrum Conforms
Specific optical rotation +6° to +8° (20°C, 589 nm) (c=2, CHCl3)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR