Stampa…

Evans blue

Fornitore: Thermo Scientific
Pericolo

195550050 195550250
ACRO195550050EA 33.9 EUR
ACRO195550050 ACRO195550250
Evans blue
Evans blue
Formula: C₃₄H₂₈N₆O₁₄S₄
Peso molecolare: 960,79 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004021
CAS Number: 314-13-6

Visualizza i codici

Risultati del test di specificazione

Appearance Dark brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 %
UV Lambda max 605 to 611 nm
Specific extinction E (1%/1cm) ≥710 (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR