Ethyl diethoxyacetate 98%
Fornitore: Thermo Fisher Scientific
Formula:
C₈H₁₆O₄ Punto di ebollizione: 198…200 °C (761 mmHg) Densità: 0,989 g/cm³ Flash Pt: 85 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00009231 CAS Number: 6065-82-3 |
Risultati del test di specificazione
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4100 to 1.4120 (20°C, 589 nm) |
GC | ≥97.5 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Sulfur compounds (as S) | ≤0.005 % |
Water soluble titr. acid | ≤0.0003 meg/g |
Thiophene | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...