Stampa…

Sodium methyl sulphate 99% pract.

Fornitore: Thermo Scientific
Attenzione

Synonyms: Methanesulphuric acid sodium salt, Methylsulphuric acid sodium salt

414900250 414901000
ACRO414900250EA 115 EUR
ACRO414900250 ACRO414901000
Sodium methyl sulphate 99% pract.
Sodium methyl sulphate
Formula: CH₃NaO₄S
Peso molecolare: 134,09 g/mol
Punto di fusione: 210 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013457
CAS Number: 512-42-5
ADR: 6.1,II

Visualizza i codici

Risultati del test di specificazione

Appearance White Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR