Stampa…
Methylboronic acid 97%
Fornitore: Thermo Fisher Scientific
Attenzione
344670010 344670050
ACRO344670010EA 46.3 EUR
ACRO344670010 ACRO344670050
Methylboronic acid 97%
Methylboronic acid
Formula: CH₅BO₂
Peso molecolare: 59.86 g/mol
Punto di fusione: 88…90 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002105
CAS Number: 13061-96-6
Visualizza i codici
Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 87 to 91 °C
Titration with NaOH 96.0 to 104.0 %