Stampa…
Allylmagnesium bromide 1 M in diethyl ether, AcroSeal®
Fornitore: Thermo Scientific
Pericolo
209531000 209538000
209531000.EA 84 EUR
209531000. 209538000.
Allylmagnesium bromide 1 M in diethyl ether, AcroSeal®
Allylmagnesium bromide
Appearance: Yellow-green to grey Solution
Formula: C₃H₅BrMg
Peso molecolare: 145,28 g/mol
Densità: 0,851 g/cm³
Flash Pt: –40 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000044
CAS Number: 1730-25-2
UN: 3399
ADR: 4.3,I
Visualizza i codici
Appearance Yellow-green to grey Solution
Titration Grignard 0.9 to 1.1 M