Stampa…
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride 97%
Fornitore: Thermo Scientific
Pericolo
Synonyms: 4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride, DMTMM, 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium chloride
348960050 348960250
ACRO348960050EA 151 EUR
ACRO348960050 ACRO348960250
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride 97%
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride
CAS No.: 3945-69-5
Formula: C₁₀H₁₇ClN₄O₃
Peso molecolare: 276,72 g/mol
Punto di fusione: 118…120 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD03613550
CAS Number: 3945-69-5
UN: 3261
ADR: 8,II
Visualizza i codici
Appearance White to almost white Powder and/or chunks
Infrared spectrum Conforms
Water ≤17 % (K.F.)
Titration with HClO4 ≥96.0 % (on dry substance)