Stampa…

Methyl orotate 98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: Methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate

208780250 208781000
ACRO208780250EA 249 EUR
ACRO208780250 ACRO208781000
Methyl orotate 98%
Methyl orotate

CAS No.: 6153-44-2

Formula: C₆H₆N₂O₄
Peso molecolare: 170,124 g/mol
Punto di fusione: 244 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00010564
CAS Number: 6153-44-2

Visualizza i codici

Risultati del test di specificazione

Appearance Off-white to yellow to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR