Stampa…
3-Chloropropylamine hydrochloride 98%
Fornitore: Thermo Fisher Scientific
Attenzione
Synonyms: 3-Chloropropan-1-amine hydrochloride
162010500 162012500 162010010
ACRO162010500EA 39.6 EUR
ACRO162010500 ACRO162012500 ACRO162010010
3-Chloropropylamine hydrochloride 98%
3-Chloropropylamine hydrochloride
CAS No.: 6276-54-6
Formula: C₃H₈ClN
Peso molecolare: 130.01 g/mol
Punto di fusione: 148…150 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012913
CAS Number: 6276-54-6
Visualizza i codici
Appearance Off-white to yellow to light beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98 % (Ionised Chloride)
Water ≤0.5 %