Stampa…

Glyoxalbis(2-hydroxyanil) 97%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 2,2'-Ethanediylidenedinitrilodiphenol, 2,2'-(Ethane-1,2-diylidenebis(azanylylidene))diphenol, Glyoxalbis(2-hydroxyanil), 1,2-Bis(2-hydroxyphenylimino)ethane

120160100 120160500
ACRO120160100EA 74.7 EUR
ACRO120160100 ACRO120160500
Glyoxalbis(2-hydroxyanil) 97%
Glyoxalbis(2-hydroxyanil)

CAS No.: 1149-16-2

Formula: C₁₄H₁₂N₂O₂
Peso molecolare: 240,26 g/mol
Punto di fusione: 201…205 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002183
CAS Number: 1149-16-2

Visualizza i codici

Risultati del test di specificazione

Appearance Beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
Sulfated ash ≤0.1 % (800°C)
Solubility (0.5 % in methanol) Clear
HPLC ≥96.0 %
Suitability (for Ca determination) Passes test
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR