Stampa…

Europium(III) thenoyltrifluoroacetonate trihydrate 95%

Fornitore: Thermo Scientific

423190010 423190050
ACRO423190010EA 114 EUR
ACRO423190010 ACRO423190050
Europium(III) thenoyltrifluoroacetonate trihydrate 95%
Europium(III) thenoyltrifluoroacetonate trihydrate

CAS No.: 21392-96-1

Formula: C₂₄H₂₁EuF₉O₉S₃
Punto di fusione: 142…147 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00150912
CAS Number: 21392-96-1

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 142 to 147 °C
Elemental Analysis ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR