Mineral oil (petroleum), puro
Fornitore: Thermo Fisher Scientific
Synonyms:
Petrolatum, liquid, Mineral oil, Paraffin oil, Nujol
Punto di ebollizione:
300 °C (1013 hPa) Densità: 0,877 g/cm³ Flash Pt: 220 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00131611 CAS Number: 8042-47-5 |
Risultati del test di specificazione
Appearance | Clear colorless Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4730 to 1.4800 (20°C, 589 nm) |
Density | (15°C) 0.860 to 0.880 g/ml |
Viscosity | 63 to 80 mm²/s (40°C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...