Oxypurinol 99%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 1H-Pyrazolo[3,4-d]pyrimidine-4,6-diol
339940010
339940050
339940250
ACRO339940010EA
74.3
EUR
ACRO339940010
ACRO339940050
ACRO339940250
Oxypurinol 99%
Oxypurinol
Appearance: Light yellow Powder
Formula:
C₅H₄N₄O₂ Peso molecolare: 152,11 g/mol Punto di fusione: >300 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00047205 CAS Number: 2465-59-0 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...