Stampa…
L-Leucinamide hydrochloride 99%
Fornitore: Thermo Scientific
Synonyms: H-Leu-NH₂·HCl, H-Leu-NH2.HCl
302980050 302980010
ACRO302980050EA 72.1 EUR
ACRO302980050 ACRO302980010
L-Leucinamide hydrochloride 99%
L-Leucinamide hydrochloride
CAS No.: 10466-61-2
Formula: C₆H₁₄N₂O
Peso molecolare: 166,65 g/mol
Punto di fusione: 244 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013012
CAS Number: 10466-61-2
Visualizza i codici
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Specific optical rotation +9° to +11° (20°C, 589 nm) (c=1, H2O)