Trioxsalen 97.5%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
4,5',8-Trimethylpsoralen, Trisoralen, TMP, 2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one
CAS No.: 3902-71-4
Formula:
C₁₄H₁₂O₃ Punto di fusione: 230…234 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00005010 CAS Number: 3902-71-4 UN: 1759 ADR: 8,I |
Risultati del test di specificazione
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 230 to 237 °C |
Loss on drying | ≤0.5 % (105 °C, 6 hrs) |
HPLC | ≥96.5 % (on dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...