N-Acetylneuraminic acid 99%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Neu5Ac, (4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid, NANA, Sialic acid, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid, Lactaminic acid
227040250
227041000
ACRO227040250EA
37.5
EUR
ACRO227040250
ACRO227041000
N-Acetylneuraminic acid 99%
N-Acetylneuraminic acid
Appearance: White to off-white Powder
Formula:
C₁₁H₁₉NO₉ Peso molecolare: 309,27 g/mol Punto di fusione: 184…186 °C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00006620 CAS Number: 131-48-6 |
Risultati del test di specificazione
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Specific optical rotation | -30° to -34° (20°C, 589 nm) (c=2, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...