Stampa…
Chromatography cartridges, Protein A/G, Pierce™
Fornitore: Thermo Fisher Scientific
89930 89931
PIER89930EA 348 EUR
PIER89930 PIER89931
Chromatography cartridges, Protein A/G, Pierce™
Colonne cromatografiche
Pierce™ Protein A/G chromatography cartridges are convenient, ready to use prepacked devices for isolation and purification of most immunoglobins.
  • Immobilised recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables polyclonal IgG purification from nearly any mammalian species
  • Resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior manufacturing method immobilises Protein A/G by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Binding capacity of >7 mg human IgG/ml resin
Pierce™ Protein A/G chromatography cartridges are compatible with the major automated liquid-chromatography systems or manual syringe processing. The cartridges attach directly to ÄKTA™ or FPLC Systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-Lok® syringe fittings or ¹/₁₆" tubing. These cartridges enable fast, easy and reproducible chromatographic separations and can be regenerated for multiple uses.
Informazioni per l'ordine: Kit includes an accessory pack (1 female Luer-Lok® adapter, 1 connector fitting, 1 column plug and bottom caps).