Stampa…
Protein L magnetic beads, Pierce™
Fornitore: Thermo Fisher Scientific
88850 88849
PIER88850EA 912 EUR
PIER88850 PIER88849
Protein L magnetic beads, Pierce™
Sferette Sferette magnetiche
These protein L magnetic beads are high-capacity and high-throughput affinity particles for antibody purification using manual or robotic magnetic separators.
  • Immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Low non-specific binding of stable, pre-blocked beads provide clean purification of antibody
  • Beads are compatible with manual and automated workflows making it versatile
Recombinant Protein L, covalently attached to a blocked magnetic bead surface, selectively binds mouse andMore Product Information