Stampa…

5-Aminolevulinic acid hydrochloride ≥98%, in polvere

Fornitore: MP Biomedicals
Attenzione

Synonyms: 5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride

0210024780 0210024790 0210024791
ICNA0210024780EA 39 EUR
ICNA0210024780 ICNA0210024790 ICNA0210024791
5-Aminolevulinic acid hydrochloride ≥98%, in polvere
5-Aminolevulinic acid hydrochloride

δ-Aminolevulinic acid hydrochloride is the universal precursor of tetrapyrroles, such as chlorophyll and heme.
δ-Aminolevulinic acid hydrochloride is used in the synthesis of hemes, vitamin B12 and bacteriochlorophyll. It is used in porphyrin biosynthesis studies. It enhances chlorophyll formation and may be used as a photodynamic herbicide is a precursor of tetrapyrroles in the biosynthesis of chlorophyll and heme.
5-Aminolevulinic acid (5-ALA) is an intermediate in heme biosynthesis and is being studied as an inducing reagent for protoporphyrin IX (PPIX) dependent fluorescence diagnosis of metastatic lymph nodes. 5-ALA is used for photodynamic therapy of diseases such as Paget’s disease and HPV infection-associated cervical condylomata acuminata.


δ-Aminolevulinic acid hydrochloride is the universal precursor of tetrapyrroles, such as chlorophyll and heme.

Formula: C₅H₁₀ClNO₃
Peso molecolare: 167,59 g/mol
Punto di fusione: 150…160 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00012869
CAS Number: 5451-09-2

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity 99.13%
Melting Point 151.3 - 152.6°C
FTIR Conforms to Standard
Loss on Drying 0,48%
Residue on Ignition 0.11%
Heavy Metals ≤20 ppm
Pseudomonas aeruginosa, Salmonella, Staphylococcus aureus Absent
Bacteria Endotoxins 0.4 EU/mg
Total Aerobic Microbial Count <100 cfu/g
Total Impurities <1.0%
Any Other Single Impurity ≤0.3%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR