Stampa…

Acido silicico sale calcico NF

Fornitore: Spectrum Chemical

C1643-1KG C1643-45KGBL
SPCMC1643-1KGEA 408 EUR
SPCMC1643-1KG SPCMC1643-45KGBL
Acido silicico sale calcico NF
Acido silicico sale calcico

Calcium silicate is also known as calcium orthosilicate, it is used as an antacid and anticaking agent in food preparation. It is suitable for personal care, cosmetic and pharmaceutical applications.


Certificazioni: NF product are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Punto di fusione: 1540 °C
Densità: 2,9 g/cm³ (20 °C)
CAS Number: 1344-95-2
EINECS: 215-710-8
Merck Index: 12,01749

Visualizza i codici

Risultati del test di specificazione

Calcium oxide (CaO) 20 - 35%
Silicon Dioxide (SiO₂) 50 - 65%
Sum of Calcium oxide, silicon dioxideand loss on ignition 90.0% Min.
pH (1 to 20 suspension) 8.4 - 11.2
Ratio of silicon dioxide to calcium oxide 0.5 - 20
Loss on ignition 20.0% Max.
Lead (Pb) 10 ppm Max.
Fluoride 50 ppm Max.
Identification Passes test
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR