Brij® S2
Fornitore: Spectrum Chemical
|
Attenzione
|
Synonyms:
Polyethylene glycol octadecyl ether, Steareth-2, Brij 72, Brij S2, Polyoxyethylene (2) stearyl ether
Brij(R) S2 or Polyoxyethylene (2) Stearyl Ether are designed to emulsify and produce stable dispersions of cosmetic materials. It appears as a colorless to light yellow liquid that may become hazy or form a precipitate
Formula:
C₁₈H₃₇(OCH₂CH₂)₂OH Peso molecolare: 358,60 g/mol Densità: 0,893 g/mL (25 °C) (lett.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00074177 CAS Number: 9005-00-9 EINECS: 500-017-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...