Stampa…
Cottonseed oil NF
Fornitore: Spectrum Chemical
CO145-200LTBL CO145-20LT CO145-4LTGL CO145-500MLGL
SPCMCO145-200LTBLEA 3600 EUR
SPCMCO145-200LTBL SPCMCO145-20LT SPCMCO145-4LTGL SPCMCO145-500MLGL
Cottonseed oil NF
Cottonseed oil
Densità: 0,91 g/cm³ (20 °C)
MDL Number: MFCD00130872
CAS Number: 8001-29-4
EINECS: 232-280-7