2-Indanylboronic acid pinacol ester 95%
Fornitore: Thermo Fisher Scientific
Synonyms:
2-(2,3-Dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity: 95%
CAS No.: 608534-44-7
Molecular Formula: C15H21BO2
Formula Weight: 244.14
Refractive Index: 1.5040
MDL No.: MFCD03788726
Formula:
C₁₅H₂₁BO₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03788726 CAS Number: 608534-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...