Stampa…

2-Thiopheneacetamide 98%

Fornitore: Thermo Scientific
Pericolo

Synonyms: Thiophene-2-acetamide

L19221.22 L19221.14 L19221.06
L19221.22EA 233 EUR
L19221.22 L19221.14 L19221.06
2-Thiopheneacetamide 98%
2-Thiopheneacetamide

Purity: 98% CAS No.: 4461-29-4 Molecular Formula: C6H7NOS Formula Weight: 141.19 Melting Point: 145-148° Beilstein Reference: 114558 MDL No.: MFCD00017556

Formula: C₆H₇NOS
Peso molecolare: 141,19 g/mol
Punto di fusione: 145…148 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00017556
CAS Number: 4461-29-4
EINECS: 224-714-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR